missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-D-glucosamine, 96.0-102.0%, Spectrum™ Chemical
$232.53 - $20966.77
Chemical Identifiers
| CAS | 7512-17-6 |
|---|---|
| Molecular Formula | C8H15NO6 |
| Molecular Weight (g/mol) | 221.21 |
| MDL Number | MFCD00061615 |
| InChI Key | OVRNDRQMDRJTHS-WZWLWIOANA-N |
| IUPAC Name | N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide |
| SMILES | CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18611890
|
Spectrum Chemical Mfg Cor
A126925GM |
25 g | Poly Bottle |
Each for $232.53
|
|
||||
|
18611892
|
Spectrum Chemical Mfg Cor
A1269100GM |
100 g | Poly Bottle |
Each for $1,163.20
|
|
||||
|
18611894
|
Spectrum Chemical Mfg Cor
A1269500GM |
500 g | Poly Bottle |
Each for $2,324.63
|
|
||||
|
18611891
|
Spectrum Chemical Mfg Cor
A12695KG |
5 kg | Poly Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18611893
|
Spectrum Chemical Mfg Cor
A126925KGBL |
25 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 7512-17-6 | |
| 221.21 | |
| OVRNDRQMDRJTHS-WZWLWIOANA-N | |
| CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O |
| C8H15NO6 | |
| MFCD00061615 | |
| N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide |
Specifications
| 7512-17-6 | |
| 6.0 to 8.0 | |
| C8H15NO6 | |
| 25 g | |
| OVRNDRQMDRJTHS-WZWLWIOANA-N | |
| N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide | |
| 96 to 102.0% |
| 1 | |
| Poly Bottle | |
| MFCD00061615 | |
| +39.0° to +43.0° | |
| CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O | |
| 221.21 | |
| Reagent |
CAUTION: For manufacturing, laboratory or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.