missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-D-glucosamine, 96.0-102.0%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor A126925KGBL
Specifications
| 7512-17-6 | |
| 6.0 to 8.0 | |
| C8H15NO6 | |
| 25 kg | |
| OVRNDRQMDRJTHS-WZWLWIOANA-N | |
| N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide | |
| 96 to 102.0% |
| 1 | |
| Fiber Drum | |
| MFCD00061615 | |
| +39.0° to +43.0° | |
| CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O | |
| 221.21 | |
| Reagent |
Chemical Identifiers
| 7512-17-6 | |
| 221.21 | |
| OVRNDRQMDRJTHS-WZWLWIOANA-N | |
| CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O |
| C8H15NO6 | |
| MFCD00061615 | |
| N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide |
CAUTION: For manufacturing, laboratory or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.