missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-D-galactosamine

CAS: 1811-31-0 | C8H15NO6 | 221.21 g/mol

Supplier:  Thermo Scientific Chemicals J6609506

 View more versions of this product

Catalog No. AAJ6609506


Only null left

Description

Description

N-Acetyl-D-galactosamine is the initial O-linked sugar to many serine and threonine residues in protein glycosylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-D-galactosamine is the initial O-linked sugar to many serine and threonine residues in protein glycosylation.

Solubility
Soluble in water at 50mg/ml

Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

N-Acetyl-D-galactosamine
160°C
C8H15NO6
5 g
dsstox_cid_28637, dsstox_rid_82907, dsstox_gsid_48711, n-acetyl-, a-d-galactosamine
OVRNDRQMDRJTHS-KEWYIRBNSA-N
N-[(3R,4R,5R,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
60196345
Solid
1811-31-0
White
MFCD00136044
2331340
Soluble in water at 50mg/ml
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O
221.21
221.21

Chemical Identifiers

1811-31-0
221.21
OVRNDRQMDRJTHS-KEWYIRBNSA-N
60196345
C8H15NO6
MFCD00136044
dsstox_cid_28637, dsstox_rid_82907, dsstox_gsid_48711, n-acetyl-, a-d-galactosamine
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 217-321-9

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only