missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-D-galactosamine

Catalog No. AAJ66095ME
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66095ME 500 mg
AAJ6609503 1 g
AAJ6609506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ66095ME Supplier Thermo Scientific Chemicals Supplier No. J66095ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1811-31-0 | C8H15NO6 | 221.21 g/mol

N-Acetyl-D-galactosamine is the initial O-linked sugar to many serine and threonine residues in protein glycosylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-D-galactosamine is the initial O-linked sugar to many serine and threonine residues in protein glycosylation.

Solubility
Soluble in water at 50mg/ml

Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1811-31-0
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.21
MDL Number MFCD00136044
InChI Key OVRNDRQMDRJTHS-KEWYIRBNSA-N
Synonym dsstox_cid_28637, dsstox_rid_82907, dsstox_gsid_48711, n-acetyl-, a-d-galactosamine
PubChem CID 60196345
SMILES CC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O

Specifications

Melting Point 160°C
Color White
Quantity 500 mg
Beilstein 2331340
Solubility Information Soluble in water at 50mg/ml
IUPAC Name N-[(2R,3S,4S,5S)-3,4,5,6-tetrahydroxy-1-oxohexan-2-yl]acetamide
Formula Weight 221.21
Physical Form Solid
Chemical Name or Material N-Acetyl-D-galactosamine
EINECSNumber 217-321-9
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only