missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-4-hydroxy-L-proline (cis- and trans- mixture) 97.0+%, TCI America™

Catalog No. p-7117813
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
A22655G 5 g
A226525G 25 g
2 options

Catalog No. A22655G

Supplier: TCI America A22655G

Only null left

Chemical Identifiers

CAS 926905-71-7
Molecular Formula C7H11NO4
Molecular Weight (g/mol) 173.17
MDL Number MFCD08166683
InChI Key BAPRUDZDYCKSOQ-UHFFFAOYNA-N
Synonym N-Acetyl-4-hydroxypyrrolidine-2-carboxylic Acid, Ac-Hyp-OH
PubChem CID 2729005
IUPAC Name 1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid
SMILES CC(=O)N1CC(O)CC1C(O)=O

Specifications

Melting Point 131°C
Quantity 5 g
Formula Weight 173.17
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-4-hydroxy-L-proline (cis- and trans- mixture)
EINECSNumber (9)-1629
TSCA No