accessibility menu, dialog, popup

UserName

CAS RN 926905-71-7

CH3ONOHOOH

CAS RN 926905-71-7

IUPAC Name: 1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid
Synonyms: 1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 173.17
Molecular Formula: C7H11NO4
InChi Key: BAPRUDZDYCKSOQ-UHFFFAOYNA-N
SMILES: CC(=O)N1CC(O)CC1C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results