missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-3,5-dimethyl-1-adamantanamine 97.0+%, TCI America™
$400.44 - $1413.08
Chemical Identifiers
| CAS | 19982-07-1 |
|---|---|
| Molecular Formula | C14H23NO |
| Molecular Weight (g/mol) | 221.344 |
| MDL Number | MFCD06656139 |
| InChI Key | WVIRSYCDAYUOMJ-UHFFFAOYSA-N |
| Synonym | 1-Acetamido-3,5-dimethyladamantane, N-Acetylmemantine, N-(3,5-Dimethyl-1-adamantyl)acetamide |
| PubChem CID | 5276209 |
| IUPAC Name | N-(3,5-dimethyl-1-adamantyl)acetamide |
| SMILES | CC(=O)NC12CC3CC(C1)(CC(C3)(C2)C)C |
Chemical Identifiers
| 19982-07-1 | |
| 221.344 | |
| WVIRSYCDAYUOMJ-UHFFFAOYSA-N | |
| 5276209 | |
| CC(=O)NC12CC3CC(C1)(CC(C3)(C2)C)C |
| C14H23NO | |
| MFCD06656139 | |
| 1-Acetamido-3,5-dimethyladamantane, N-Acetylmemantine, N-(3,5-Dimethyl-1-adamantyl)acetamide | |
| N-(3,5-dimethyl-1-adamantyl)acetamide |
Specifications
| 19982-07-1 | |
| White-Yellow | |
| MFCD06656139 | |
| 1-Acetamido-3,5-dimethyladamantane, N-Acetylmemantine, N-(3,5-Dimethyl-1-adamantyl)acetamide | |
| CC(=O)NC12CC3CC(C1)(CC(C3)(C2)C)C | |
| 221.344 | |
| 221.34 | |
| Crystalline Powder |
| 112°C | |
| C14H23NO | |
| 1 g | |
| WVIRSYCDAYUOMJ-UHFFFAOYSA-N | |
| N-(3,5-dimethyl-1-adamantyl)acetamide | |
| 5276209 | |
| ≥97.0% (GC) | |
| N-Acetyl-3,5-dimethyl-1-adamantanamine |
Safety and Handling
TSCA : No