accessibility menu, dialog, popup

UserName

CH3ONHCH3CH3

CAS RN 19982-07-1

IUPAC Name: N-(3,5-dimethyladamantan-1-yl)acetamide
Synonyms: N-(3,5-dimethyladamantan-1-yl)acetamide
Molecular Weight (g/mol): 221.34
Molecular Formula: C14H23NO
InChi Key: WVIRSYCDAYUOMJ-UHFFFAOYNA-N
SMILES: CC(=O)NC12CC3CC(C)(CC(C)(C3)C1)C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results