missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-3,5-dimethyl-1-adamantanamine 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 112°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 221.34 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-Acetyl-3,5-dimethyl-1-adamantanamine |
Safety and Handling
| TSCA | No |