missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetoacetylmorpholine 98.0+%, TCI America™
Supplier: TCI America A072625G
Specifications
| N-Acetoacetylmorpholine | |
| 68°C | |
| 140°C | |
| MFCD00059774 | |
| RKPILPRSNWEZJV-UHFFFAOYSA-N | |
| 1-(morpholin-4-yl)butane-1,3-dione | |
| 85550 | |
| ≥98.0% (HPLC,N) |
| 16695-54-8 | |
| White | |
| C8H13NO3 | |
| 25 g | |
| CC(=O)CC(=O)N1CCOCC1 | |
| 171.20 | |
| 171.20 | |
| Crystalline Powder |
Chemical Identifiers
| 16695-54-8 | |
| 171.20 | |
| RKPILPRSNWEZJV-UHFFFAOYSA-N | |
| 1-(morpholin-4-yl)butane-1,3-dione |
| C8H13NO3 | |
| MFCD00059774 | |
| 85550 | |
| CC(=O)CC(=O)N1CCOCC1 |
Safety and Handling
EINECSNumber : (9)-1635
TSCA : No