accessibility menu, dialog, popup

UserName

CAS RN 16695-54-8

H3COONO

CAS RN 16695-54-8

IUPAC Name: 1-(morpholin-4-yl)butane-1,3-dione
Synonyms: 1-(morpholin-4-yl)butane-1,3-dione
Molecular Weight (g/mol): 171.2
Molecular Formula: C8H13NO3
InChi Key: RKPILPRSNWEZJV-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)N1CCOCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results