missing translation for 'onlineSavingsMsg'
Learn More

N-[(9S)-8alpha-Cinchonan-9-yl]quinoline-8-sulfonamide 98.0+%, TCI America™

Catalog No. c2728100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2728100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2728100MG Supplier TCI America Supplier No. C2728100MG
Only null left

Chemical Identifiers

CAS 1440939-88-7
Molecular Formula C28H28N4O2S
Molecular Weight (g/mol) 484.618
InChI Key RNPQJTUCXSVJMB-JPDZPCQLSA-N
Synonym (S)-N-8-Quinolinesulfonyl-(quinolin-4-yl)(8-vinylquinuclidin-2-yl)methanamine
PubChem CID 91972085
IUPAC Name N-[(S)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-quinolin-4-ylmethyl]quinoline-8-sulfonamide
SMILES C=CC1CN2CCC1CC2C(C3=CC=NC4=CC=CC=C34)NS(=O)(=O)C5=CC=CC6=C5N=CC=C6

Specifications

Melting Point 260°C
Color Yellow
Quantity 100 mg
Formula Weight 484.62
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9S)-8alpha-Cinchonan-9-yl]quinoline-8-sulfonamide