accessibility menu, dialog, popup

UserName

CAS RN 1440939-88-7

IUPAC Name: N-({5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl}(quinolin-4-yl)methyl)quinoline-8-sulfonamide
Synonyms: N-({5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl}(quinolin-4-yl)methyl)quinoline-8-sulfonamide
Molecular Weight (g/mol): 484.62
Molecular Formula: C28H28N4O2S
InChi Key: RNPQJTUCXSVJMB-UHFFFAOYNA-N
SMILES: C=CC1CN2CCC1CC2C(NS(=O)(=O)C1=C2N=CC=CC2=CC=C1)C1=C2C=CC=CC2=NC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results