missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine 98.0+%, TCI America™

Catalog No. p-7129811
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06525G 5 g
F065225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06525G Supplier TCI America Supplier No. F06525G
Only null left

Chemical Identifiers

CAS 103213-32-7
Molecular Formula C37H30NO4S
Molecular Weight (g/mol) 584.71
MDL Number MFCD00038538
InChI Key KLBPUVPNPAJWHZ-UMSFTDKQSA-M
Synonym fmoc-cys trt-oh, fmoc-s-trityl-l-cysteine, fmoc-l-cys trt-oh, n-fmoc-s-trityl-l-cysteine, fmoc-cys trt, n alpha-fluorenylmethyloxycarbonyl-s-tritylcysteine, r-9h-fluoren-9-yl methyl 1-hydroxy-3-tritylthio propan-2-yl carbamate, fmoc-s-trityl-cys, n-9h-fluoren-9-ylmethoxy carbonyl-s-triphenylmethyl-l-cysteine, l-cysteine, n-9h-fluoren-9-ylmethoxy carbonyl-s-triphenylmethyl
PubChem CID 128239
IUPAC Name (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[(triphenylmethyl)sulfanyl]propanoate
SMILES [O-]C(=O)[C@H](CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 175°C
Color White-Yellow
Quantity 5 g
Formula Weight 585.72
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine