missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(tert-butyl)-L-cysteine 98.0+%, TCI America™

Catalog No. p-7130004
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09721G 1 g
F09725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09721G Supplier TCI America Supplier No. F09721G
Only null left

Chemical Identifiers

CAS 67436-13-9
Molecular Formula C22H25NO4S
Molecular Weight (g/mol) 399.505
MDL Number MFCD00037130
InChI Key IXAYZHCPEYTWHW-IBGZPJMESA-N
Synonym fmoc-cys tbu-oh, fmoc-s-tert-butyl-l-cysteine, fmoc-s-t-butyl-l-cysteine, n-fmoc-s-tert-butyl-l-cysteine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butylthio propanoic acid, fmoc-r-2-amino-3-s-t-butylthio butanoic acid, 2r-3-tert-butylsulfanyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, ambotzfaa1716, pubchem19015, fmoc-cys but-oh
PubChem CID 7009544
IUPAC Name (2R)-3-tert-butylsulfanyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES CC(C)(C)SCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 138°C
Color White
Quantity 1 g
Formula Weight 399.51
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(tert-butyl)-L-cysteine
TSCA No
Recommended Storage Refrigerator