missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-D-threonine 98.0+%, TCI America™
$95.06 - $277.75
Chemical Identifiers
| CAS | 138797-71-4 |
|---|---|
| Molecular Formula | C23H27NO5 |
| Molecular Weight (g/mol) | 397.47 |
| MDL Number | MFCD00080292 |
| InChI Key | LZOLWEQBVPVDPR-VBKZILBWSA-N |
| Synonym | fmoc-d-thr tbu-oh, fmoc-o-tert-butyl-d-threonine, fmoc-d-thr but-oh, fmoc-thr but-oh, 2r,3s-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxy butanoic acid, fmoc-o-tert.butyl-d-threonine, ambotzfaa1509, pubchem10044, n-fmoc-o-tert-butyl-d-threonine |
| PubChem CID | 7021418 |
| IUPAC Name | (2R,3S)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid |
| SMILES | C[C@H](OC(C)(C)C)[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O |
Chemical Identifiers
| 138797-71-4 | |
| 397.47 | |
| LZOLWEQBVPVDPR-VBKZILBWSA-N | |
| 7021418 | |
| C[C@H](OC(C)(C)C)[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O |
| C23H27NO5 | |
| MFCD00080292 | |
| fmoc-d-thr tbu-oh, fmoc-o-tert-butyl-d-threonine, fmoc-d-thr but-oh, fmoc-thr but-oh, 2r,3s-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxy butanoic acid, fmoc-o-tert.butyl-d-threonine, ambotzfaa1509, pubchem10044, n-fmoc-o-tert-butyl-d-threonine | |
| (2R,3S)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid |
Specifications
| 138797-71-4 | |
| White-Yellow | |
| MFCD00080292 | |
| fmoc-d-thr tbu-oh, fmoc-o-tert-butyl-d-threonine, fmoc-d-thr but-oh, fmoc-thr but-oh, 2r,3s-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxy butanoic acid, fmoc-o-tert.butyl-d-threonine, ambotzfaa1509, pubchem10044, n-fmoc-o-tert-butyl-d-threonine | |
| C[C@H](OC(C)(C)C)[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O | |
| 397.47 | |
| 397.47 | |
| Crystalline Powder |
| 133°C | |
| C23H27NO5 | |
| 1 g | |
| LZOLWEQBVPVDPR-VBKZILBWSA-N | |
| (2R,3S)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid | |
| 7021418 | |
| ≥98.0% (HPLC,T) | |
| N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-D-threonine |
Safety and Handling
TSCA : No