missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-norvaline 98.0+%, TCI America™

Catalog No. F11171G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11171G 1 g
F11175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F11171G Supplier TCI America Supplier No. F11171G

Chemical Identifiers

CAS 135112-28-6
Molecular Formula C20H21NO4
Molecular Weight (g/mol) 339.39
MDL Number MFCD00155631
InChI Key JBIJSEUVWWLFGV-SFHVURJKSA-N
Synonym fmoc-nva-oh, fmoc-l-norvaline, n-9-fluorenylmethyloxycarbonyl-l-norvaline, n-fmoc-l-norvaline, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, l-norvaline, n-9h-fluoren-9-ylmethoxy carbonyl, fmoc-norvaline, fmoc-l-nva-oh, ambotzfaa1415, pubchem14961
PubChem CID 7016885
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid
SMILES CCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 155°C
Color White
Quantity 1 g
Formula Weight 339.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-norvaline
TSCA No
Recommended Storage Refrigerator