missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-methionine 98.0+%, TCI America™
$26.84 - $113.71
Chemical Identifiers
| CAS | 71989-28-1 |
|---|---|
| Molecular Formula | C20H21NO4S |
| Molecular Weight (g/mol) | 371.45 |
| MDL Number | MFCD00037134 |
| InChI Key | BUBGAUHBELNDEW-SFHVURJKSA-N |
| Synonym | fmoc-met-oh, fmoc-l-methionine, n-fmoc-l-methionine, fmoc-l-met-oh, n-9h-fluoren-9-ylmethoxy carbonyl-l-methionine, fmoc-met, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylsulfanyl butanoic acid, 9-fluorenylmethoxycarbonyl-l-methionine, methionine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-l-methionine |
| PubChem CID | 2724632 |
| IUPAC Name | (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-(methylsulfanyl)butanoic acid |
| SMILES | CSCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
Chemical Identifiers
| 71989-28-1 | |
| 371.45 | |
| BUBGAUHBELNDEW-SFHVURJKSA-N | |
| 2724632 | |
| CSCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
| C20H21NO4S | |
| MFCD00037134 | |
| fmoc-met-oh, fmoc-l-methionine, n-fmoc-l-methionine, fmoc-l-met-oh, n-9h-fluoren-9-ylmethoxy carbonyl-l-methionine, fmoc-met, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylsulfanyl butanoic acid, 9-fluorenylmethoxycarbonyl-l-methionine, methionine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-l-methionine | |
| (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-(methylsulfanyl)butanoic acid |
Specifications
| 71989-28-1 | |
| White | |
| MFCD00037134 | |
| fmoc-met-oh, fmoc-l-methionine, n-fmoc-l-methionine, fmoc-l-met-oh, n-9h-fluoren-9-ylmethoxy carbonyl-l-methionine, fmoc-met, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylsulfanyl butanoic acid, 9-fluorenylmethoxycarbonyl-l-methionine, methionine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-l-methionine | |
| CSCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
| 371.45 | |
| 371.45 | |
| Crystalline Powder |
| 141°C | |
| C20H21NO4S | |
| 1 g | |
| BUBGAUHBELNDEW-SFHVURJKSA-N | |
| (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-(methylsulfanyl)butanoic acid | |
| 2724632 | |
| ≥98.0% (HPLC,T) | |
| N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-methionine |
Safety and Handling
TSCA : No