missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-methionine 98.0+%, TCI America™

Supplier:  TCI America F029625G

 View more versions of this product

Catalog No. F029625G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-methionine
141°C
C20H21NO4S
25 g
BUBGAUHBELNDEW-SFHVURJKSA-N
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-(methylsulfanyl)butanoic acid
2724632
≥98.0% (HPLC,T)
71989-28-1
White
MFCD00037134
fmoc-met-oh, fmoc-l-methionine, n-fmoc-l-methionine, fmoc-l-met-oh, n-9h-fluoren-9-ylmethoxy carbonyl-l-methionine, fmoc-met, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylsulfanyl butanoic acid, 9-fluorenylmethoxycarbonyl-l-methionine, methionine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-l-methionine
CSCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
371.45
371.45
Crystalline Powder

Chemical Identifiers

71989-28-1
371.45
BUBGAUHBELNDEW-SFHVURJKSA-N
2724632
CSCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
C20H21NO4S
MFCD00037134
fmoc-met-oh, fmoc-l-methionine, n-fmoc-l-methionine, fmoc-l-met-oh, n-9h-fluoren-9-ylmethoxy carbonyl-l-methionine, fmoc-met, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylsulfanyl butanoic acid, 9-fluorenylmethoxycarbonyl-l-methionine, methionine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-l-methionine
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-(methylsulfanyl)butanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions