missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-homophenylalanine 98.0+%, TCI America™

Catalog No. F10251G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10251G Supplier TCI America Supplier No. F10251G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 132684-59-4
Melting Point 143°C
Color White
Molecular Formula C25H23NO4
Quantity 1 g
MDL Number MFCD00077046
Synonym fmoc-l-homophenylalanine, fmoc-homophe-oh, fmoc-hophe-oh, fmoc-l-homophenyl-ala, fmoc-l-homophe, fmoc-l-hphe-oh, n-fmoc-l-homophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-phenylbutanoic acid, s-2-fmoc-amino-4-phenylbutyric acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-phenylbutanoic acid
InChI Key CIHPCIUGLIZADU-QHCPKHFHSA-N
SMILES OC(=O)[C@H](CCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-phenylbutanoic acid
Molecular Weight (g/mol) 401.46
PubChem CID 2761467
Formula Weight 401.46
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-homophenylalanine
Show More Show Less
TSCA No
Recommended Storage Refrigerator