missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-2-phenylglycine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 176°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 373.41 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-2-phenylglycine |
Safety and Handling
| TSCA | No |