missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-valine 98.0+%, TCI America™

Catalog No. p-7129783
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06105G 5 g
F061025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06105G Supplier TCI America Supplier No. F06105G
Only null left

Chemical Identifiers

CAS 84624-17-9
Molecular Formula C20H21NO4
Molecular Weight (g/mol) 339.39
MDL Number MFCD00062953
InChI Key UGNIYGNGCNXHTR-GOSISDBHSA-N
Synonym fmoc-d-val-oh, fmoc-d-valine, n-fmoc-d-valine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylbutanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-d-valine, n-9-fluorenylmethoxycarbonyl-d-valine, 9-fluorenylmethoxycarbonyl-d-valine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-methylbutanoic acid, n-9-fluorenylmethyloxycarbonyl-d-valine, 9h-fluoren-9-yl methoxy carbonyl-d-valine
PubChem CID 1549479
IUPAC Name (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylbutanoic acid
SMILES CC(C)[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 146°C
Color White
Quantity 5 g
Formula Weight 339.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-valine