missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-threonine 98.0+%, TCI America™

Catalog No. F06081G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06081G 1 g
F06085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06081G Supplier TCI America Supplier No. F06081G
Only null left

Chemical Identifiers

CAS 157355-81-2
Molecular Formula C19H19NO5
Molecular Weight (g/mol) 341.363
InChI Key OYULCCKKLJPNPU-APPDUMDISA-N
Synonym N-Fmoc-D-threonine, Fmoc-D-Thr-OH
PubChem CID 6992532
IUPAC Name (2R,3S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-hydroxybutanoic acid
SMILES CC(C(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 341.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-threonine