missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-serine 95.0+%, TCI America™

Catalog No. p-7129780
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06071G 1 g
F06075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06071G Supplier TCI America Supplier No. F06071G
Only null left

Chemical Identifiers

CAS 116861-26-8
Molecular Formula C18H17NO5
Molecular Weight (g/mol) 327.336
MDL Number MFCD00077068
InChI Key JZTKZVJMSCONAK-MRXNPFEDSA-N
Synonym fmoc-d-ser-oh, fmoc-d-serine, n-9h-fluoren-9-ylmethoxy carbonyl-d-serine, n-fmoc-d-serine, d-serine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-serine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-hydroxypropanoic acid, ambotzfaa1508, pubchem10038, n-fmoc-d-ala oh
PubChem CID 688637
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-hydroxypropanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CO)C(=O)O

Specifications

Melting Point 112°C
Color White-Yellow
Quantity 1 g
Formula Weight 327.34
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-serine