missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine Hydrate 98.0+%, TCI America™

Catalog No. p-7129775
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05965G 5 g
F059625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05965G Supplier TCI America Supplier No. F05965G
Only null left

Chemical Identifiers

CAS 79990-15-1
Molecular Formula C18H17NO4
Molecular Weight (g/mol) 311.34
MDL Number MFCD00062960
InChI Key QWXZOFZKSQXPDC-UHFFFAOYNA-N
Synonym fmoc-d-ala-oh, fmoc-d-alanine, n-fmoc-d-alanine, n-9-fluorenylmethoxycarbonyl-d-alanine, d-alanine, n-9h-fluoren-9-ylmethoxy carbonyl, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, n-9h-fluoren-9ylmethoxy carbonyl-d-alanine, r-2-9h-fluoren-9-yl-methoxycarbonylamino-propionic acid
PubChem CID 2724627
IUPAC Name 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
SMILES CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 157°C
Color Yellow
Quantity 5 g
Formula Weight 311.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine Hydrate