missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-beta-alanine 99.0+%, TCI America™
Supplier: TCI America F088225G
Specifications
| N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-beta-alanine | |
| 148°C | |
| C18H16NO4 | |
| 25 g | |
| LINBWYYLPWJQHE-UHFFFAOYSA-M | |
| 3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoate | |
| 2724630 | |
| ≥99.0% (HPLC) |
| 35737-10-1 | |
| White | |
| MFCD00063328 | |
| fmoc-beta-alanine, fmoc-beta-ala-oh, fmoc-beta-ala-oh, fmoc-b-ala-oh, n-fmoc-beta-alanine, fmoc-b-alanine, 3-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-, fmoc-, 3-9h-fluoren-9-yl methoxy carbonyl amino propanoic acid, 3-9h-fluoren-9-yl methoxy carbonyl amino propanoic acid | |
| [O-]C(=O)CCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
| 310.33 | |
| 311.34 | |
| Crystalline Powder |
Chemical Identifiers
| 35737-10-1 | |
| 310.33 | |
| LINBWYYLPWJQHE-UHFFFAOYSA-M | |
| 2724630 | |
| [O-]C(=O)CCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
| C18H16NO4 | |
| MFCD00063328 | |
| fmoc-beta-alanine, fmoc-beta-ala-oh, fmoc-beta-ala-oh, fmoc-b-ala-oh, n-fmoc-beta-alanine, fmoc-b-alanine, 3-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-, fmoc-, 3-9h-fluoren-9-yl methoxy carbonyl amino propanoic acid, 3-9h-fluoren-9-yl methoxy carbonyl amino propanoic acid | |
| 3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator