missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-trans-(tert-butoxy)-L-proline 98.0+%, TCI America™

Catalog No. F10951G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10951G 1 g
F10955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10951G Supplier TCI America Supplier No. F10951G
Only null left

Chemical Identifiers

CAS 122996-47-8
Molecular Formula C24H27NO5
Molecular Weight (g/mol) 409.482
MDL Number MFCD00151930
InChI Key WPBXBYOKQUEIDW-FXMQYSIJSA-N
Synonym fmoc-4-tert-butoxy-l-proline, 2s-4-tert-butoxy-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, fmoc-o-tert-butyl-l-hydroxyproline, ksc491o9t, n-alpha-9-fluorenylmethyloxycarbonyl-o-t-butyl-trans-l-hydroxyproline, 2s-1-9h-fluoren-9-ylmethoxycarbonyl-4-2-methylpropan-2-yl oxy pyrrolidine-2-carboxylic acid
PubChem CID 6916162
IUPAC Name (2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-[(2-methylpropan-2-yl)oxy]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O

Specifications

Melting Point 62°C
Color White-Yellow
Quantity 1 g
Formula Weight 409.48
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-trans-(tert-butoxy)-L-proline