missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine 98.0+%, TCI America™
$59.65 - $178.95
Chemical Identifiers
| CAS | 135673-97-1 |
|---|---|
| Molecular Formula | C24H27NO4 |
| Molecular Weight (g/mol) | 393.48 |
| MDL Number | MFCD00065614 |
| InChI Key | HIJAUEZBPWTKIV-UHFFFAOYNA-N |
| Synonym | fmoc-cha-oh, fmoc-3-cyclohexyl-l-alanine, fmoc-l-cyclohexylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-cyclohexylpropanoic acid, n-fmoc-3-cyclohexyl-l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-3-cyclohexyl-l-alanine, n-alpha-9-fluorenylmethyloxycarbonyl-l-cyclohexylalanine, 2s-3-cyclohexyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2s-3-cyclohexyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-3-cyclohexylalanine |
| PubChem CID | 978326 |
| IUPAC Name | 3-cyclohexyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid |
| SMILES | OC(=O)C(CC1CCCCC1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Chemical Identifiers
| 135673-97-1 | |
| 393.48 | |
| HIJAUEZBPWTKIV-UHFFFAOYNA-N | |
| 978326 | |
| OC(=O)C(CC1CCCCC1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
| C24H27NO4 | |
| MFCD00065614 | |
| fmoc-cha-oh, fmoc-3-cyclohexyl-l-alanine, fmoc-l-cyclohexylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-cyclohexylpropanoic acid, n-fmoc-3-cyclohexyl-l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-3-cyclohexyl-l-alanine, n-alpha-9-fluorenylmethyloxycarbonyl-l-cyclohexylalanine, 2s-3-cyclohexyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2s-3-cyclohexyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-3-cyclohexylalanine | |
| 3-cyclohexyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid |
Specifications
| 135673-97-1 | |
| White | |
| MFCD00065614 | |
| fmoc-cha-oh, fmoc-3-cyclohexyl-l-alanine, fmoc-l-cyclohexylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-cyclohexylpropanoic acid, n-fmoc-3-cyclohexyl-l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-3-cyclohexyl-l-alanine, n-alpha-9-fluorenylmethyloxycarbonyl-l-cyclohexylalanine, 2s-3-cyclohexyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2s-3-cyclohexyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-3-cyclohexylalanine | |
| OC(=O)C(CC1CCCCC1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
| 393.48 | |
| 393.48 | |
| Crystalline Powder |
| 140°C | |
| C24H27NO4 | |
| 1 g | |
| HIJAUEZBPWTKIV-UHFFFAOYNA-N | |
| 3-cyclohexyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid | |
| 978326 | |
| ≥98.0% (HPLC,T) | |
| N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator