missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(4-pyridyl)-L-alanine 98.0+%, TCI America™
Supplier: TCI America F09275G
Specifications
| N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(4-pyridyl)-L-alanine | |
| 219°C | |
| C23H20N2O4 | |
| 5 g | |
| SCSSXJVRZMQUKA-NRFANRHFSA-N | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-4-ylpropanoic acid | |
| 978322 | |
| ≥98.0% (HPLC,T) |
| 169555-95-7 | |
| Yellow | |
| MFCD00672566 | |
| fmoc-l-4-pyridylalanine, fmoc-4-pal-oh, fmoc-3-4-pyridyl-l-alanine, fmoc-ala 4-pyridyl-oh, fmoc-beta-4-pyridyl-ala-oh, n-fmoc-3-4-pyridyl-l-alanine, fmoc-l-3-4-pyridyl-alanine, fmoc-4'-pyridyl-l-ala, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-4-yl propanoic acid, fmoc-3-4-pyridyl-d-alanine | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=NC=C4)C(=O)O | |
| 388.423 | |
| 388.42 | |
| Crystalline Powder |
Chemical Identifiers
| 169555-95-7 | |
| 388.423 | |
| SCSSXJVRZMQUKA-NRFANRHFSA-N | |
| 978322 | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=NC=C4)C(=O)O |
| C23H20N2O4 | |
| MFCD00672566 | |
| fmoc-l-4-pyridylalanine, fmoc-4-pal-oh, fmoc-3-4-pyridyl-l-alanine, fmoc-ala 4-pyridyl-oh, fmoc-beta-4-pyridyl-ala-oh, n-fmoc-3-4-pyridyl-l-alanine, fmoc-l-3-4-pyridyl-alanine, fmoc-4'-pyridyl-l-ala, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-4-yl propanoic acid, fmoc-3-4-pyridyl-d-alanine | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-4-ylpropanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator