missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(4-pyridyl)-L-alanine 98.0+%, TCI America™

Catalog No. F09271G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09271G 1 g
F09275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09271G Supplier TCI America Supplier No. F09271G

Chemical Identifiers

CAS 169555-95-7
Molecular Formula C23H20N2O4
Molecular Weight (g/mol) 388.423
MDL Number MFCD00672566
InChI Key SCSSXJVRZMQUKA-NRFANRHFSA-N
Synonym fmoc-l-4-pyridylalanine, fmoc-4-pal-oh, fmoc-3-4-pyridyl-l-alanine, fmoc-ala 4-pyridyl-oh, fmoc-beta-4-pyridyl-ala-oh, n-fmoc-3-4-pyridyl-l-alanine, fmoc-l-3-4-pyridyl-alanine, fmoc-4'-pyridyl-l-ala, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-4-yl propanoic acid, fmoc-3-4-pyridyl-d-alanine
PubChem CID 978322
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-4-ylpropanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=NC=C4)C(=O)O

Specifications

Melting Point 219°C
Color Yellow
Quantity 1 g
Formula Weight 388.42
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(4-pyridyl)-L-alanine
TSCA No
Recommended Storage Refrigerator