missing translation for 'onlineSavingsMsg'
Learn More

N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide Hydrochloride 98.0+%, TCI America™

Catalog No. A2409100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2409100MG 100 mg
A24091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2409100MG Supplier TCI America Supplier No. A2409100MG

Chemical Identifiers

CAS 61714-27-0
Molecular Formula C16H22Cl2N2O2S
Molecular Weight (g/mol) 377.32
MDL Number MFCD00012559
InChI Key OMMOSRLIFSCDBL-UHFFFAOYSA-N
Synonym w-7 hydrochloride, n-6-aminohexyl-5-chloro-1-naphthalenesulfonamide hydrochloride, w-7, hydrochloride, n-6-aminohexyl-5-chloronaphthalene-1-sulfonamide hydrochloride, c16h21cln2o2s hydrochloride, 1-naphthalenesulfonamide, n-6-aminohexyl-5-chloro-, monohydrochloride, n-6-aminohexyl-5-chloro-1-naphthalenesulfonamide monohydrochloride, n-6-aminohexyl-5-chloro-1-naphthalenesulfonamide hcl, n-6-aminohexyl-5-chloro-1-naphthalenesulfonamide, hcl, n-aminohexyl-5-chloro-1-napthalenesulfonamide hydrochloride
PubChem CID 124887
IUPAC Name hydrogen N-(6-aminohexyl)-5-chloronaphthalene-1-sulfonamide chloride
SMILES [H+].[Cl-].NCCCCCCNS(=O)(=O)C1=C2C=CC=C(Cl)C2=CC=C1

Specifications

Melting Point 222°C
Quantity 100 mg
Formula Weight 377.32
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide Hydrochloride
TSCA No
Recommended Storage Freezer