missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Vinylbenzyl)-N,N-dimethylamine, 90%, technical, stabilized

Catalog No. AC422550050
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC422550050 5 g Glass bottle
AC422550500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC422550050 Supplier Thermo Scientific Chemicals Supplier No. 422550050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2245-52-5 | C11H16N | 162.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2245-52-5
Molecular Formula C11H16N
Molecular Weight (g/mol) 162.26
MDL Number MFCD00078270
InChI Key XQBHAZDVLGNSOJ-UHFFFAOYSA-O
Synonym benzenemethanamine, 4-ethenyl-n,n-dimethyl, n,n-dimethyl-1-4-vinylphenyl methanamine, n-4-vinylbenzyl-n,n-dimethylamine, n,n-dimethyl-4'-vinylbenzylamine, 4-vinylbenzyldimethylamine, p-dimethylaminomethylstyrene, 4-n,n-dimethylaminomethyl styrene, 4-n,n,-dimethylaminomethyl styrene, dimethyl 4-vinylphenyl methyl amine, benzenemethanamine,4-ethenyl-n,n-dimethyl
PubChem CID 519956
IUPAC Name 1-(4-ethenylphenyl)-N,N-dimethylmethanamine
SMILES C[NH+](C)CC1=CC=C(C=C)C=C1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Yellow to Brown
Density 0.9200g/mL
Boiling Point 72°C to 74°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.5360 to 1.54
Linear Formula CH2CHC6H4CH2N(CH3)2
Quantity 5 g
Specific Gravity 0.92
Stabilizer 0.1% P-tert.-butylcatechol
Formula Weight 161.25
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material N-(4-Vinylbenzyl)-N, N-dimethylamine, tech., 90%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and k

RUO – Research Use Only