missing translation for 'onlineSavingsMsg'
Learn More

N-[4-(Trifluoromethyl)phenyl]thiourea, 98%

Catalog No. AAL1235503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1235503 1 g
AAL1235506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1235503 Supplier Thermo Scientific Chemicals Supplier No. L1235503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1736-72-7 | C8H7F3N2S | 220.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1736-72-7
Molecular Formula C8H7F3N2S
Molecular Weight (g/mol) 220.21
MDL Number MFCD00041189
InChI Key OWTDDZMFRLUBQI-UHFFFAOYSA-N
Synonym 1-4-trifluoromethyl phenyl thiourea, 4-trifluoromethyl phenyl thiourea, 1-4-trifluoromethyl phenyl-2-thiourea, 4-trifluoromethyl phenylthiourea, n-4-trifluoromethyl phenyl thiourea, 4-trifluoromethyl-phenyl-thiourea, thiourea, 4-trifluoromethyl phenyl, amino 4-trifluoromethyl phenyl amino methane-1-thione
PubChem CID 2777727
IUPAC Name [4-(trifluoromethyl)phenyl]thiourea
SMILES NC(=S)NC1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 142°C to 146°C
Odor Odorless
Quantity 1 g
UN Number UN2811
Beilstein 2695479
Formula Weight 220.22
Percent Purity 98%
Chemical Name or Material N-[4-(Trifluoromethyl)phenyl]thiourea
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only