missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Pyridylmethyl)ethylamine, 96%

Catalog No. AAL1371406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1371406 5 g
AAL1371414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1371406 Supplier Thermo Scientific Chemicals Supplier No. L1371406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33403-97-3 | C8H12N2 | 136.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33403-97-3
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00023632
InChI Key ZBAMQLFFVBPAOX-UHFFFAOYSA-N
Synonym 4-ethylaminomethyl pyridine, n-pyridin-4-ylmethyl ethanamine, n-4-pyridylmethyl ethylamine, 4-pyridinemethanamine, n-ethyl, unii-eye32x3w6x, n-ethyl-4-pyridinemethanamine, ethyl pyridin-4-ylmethyl amine, 4-ethylamino methyl pyridine, n-ethyl-4-picolylamine, eye32x3w6x
PubChem CID 96681
IUPAC Name N-(pyridin-4-ylmethyl)ethanamine
SMILES CCNCC1=CC=NC=C1

Specifications

Density 0.98
Boiling Point 103°C to 105°C (8 mmHg)
Flash Point 89°C (192°F)
Refractive Index 1.5175
Quantity 5 g
Beilstein 471685
Formula Weight 136.2
Percent Purity 96%
Chemical Name or Material N-(4-Pyridylmethyl)ethylamine
Hazard Category H314
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 251-506-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only