missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Pyridyl)pivalamide 98.0+%, TCI America™

Catalog No. P20441G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20441G 1 g
P20445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20441G Supplier TCI America Supplier No. P20441G

Chemical Identifiers

CAS 70298-89-4
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00996248
InChI Key JCMMVFHXRDNILC-UHFFFAOYSA-N
Synonym 2,2-dimethyl-n-pyridin-4-yl-propionamide, n-pyridin-4-yl pivalamide, 2,2-dimethyl-n-pyridin-4-yl propanamide, 2,2-dimethyl-n-4-pyridinyl propanamide, n-4-pyridyl pivalamide, 2,2-dimethyl-n-4-pyridyl propanamide, 4-2,2,2-trimethylacetamido pyridine, 2,2-dimethyl-n-4-pyridyl propionamide, 4-pivaloylaminopyridine, maybridge3_004407
PubChem CID 427059
IUPAC Name 2,2-dimethyl-N-pyridin-4-ylpropanamide
SMILES CC(C)(C)C(=O)NC1=CC=NC=C1

Specifications

Melting Point 133°C
Color White
Quantity 1 g
Formula Weight 178.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Pyridyl)pivalamide