missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Nitrophenyl)thiourea, 98%

Catalog No. p-7045074
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0933103 1 g
AAL0933106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0933103 Supplier Thermo Scientific Chemicals Supplier No. L0933103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3696-22-8 | C7H7N3O2S | 197.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3696-22-8
Molecular Formula C7H7N3O2S
Molecular Weight (g/mol) 197.21
MDL Number MFCD00007304
InChI Key BLYAANPIHFKKMQ-UHFFFAOYSA-N
Synonym 1-4-nitrophenyl-2-thiourea, n-4-nitrophenyl thiourea, 4-nitrophenyl thiourea, 1-4-nitrophenyl thiourea, p-nitrophenyl thiourea, thiourea, 4-nitrophenyl, n-p-nitrophenyl thiourea, acmc-1cny6, 4-nitro-phenyl-thiourea, n-4-nitrophenyl thiourea #
PubChem CID 2760234
IUPAC Name (4-nitrophenyl)thiourea
SMILES NC(=S)NC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point ∽207°C (decomposition)
Odor Odorless
Quantity 1 g
UN Number UN2811
Beilstein 2806574
Formula Weight 197.22
Percent Purity 98%
Chemical Name or Material N-(4-Nitrophenyl)thiourea
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 223-021-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only