missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Methyl-2-pyridyl)thiourea, 97%

Catalog No. p-7032253
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3364803 1 g
AAH3364806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AAH3364803 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' H3364803

CAS: 21242-21-7 | C7H9N3S | 167.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21242-21-7
Molecular Formula C7H9N3S
Molecular Weight (g/mol) 167.23
MDL Number MFCD04117348
InChI Key ALLBLAKJCNRLLS-UHFFFAOYSA-N
Synonym 4-methyl-pyridin-2-yl-thiourea, 4-methylpyridin-2-yl-thiourea, 1-4-methylpyridin-2-yl thiourea, 4-methylpyridin-2-yl thiourea, n-4-methyl-2-pyridyl thiourea, n-4-methylpyridin-2-yl thiourea, thiourea, 4-methyl-2-pyridinyl
PubChem CID 2760059
IUPAC Name (4-methylpyridin-2-yl)thiourea
SMILES CC1=CC(NC(N)=S)=NC=C1

Specifications

Melting Point 216°C to 220°C
Quantity 1 g
Formula Weight 167.23
Percent Purity 97%
Chemical Name or Material N-(4-Methyl-2-pyridyl)thiourea
missing translation for 'healthHazard1' H302
missing translation for 'healthHazard2' GHS H Statement
H301
Toxic if swallowed.
missing translation for 'healthHazard3' P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only