missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Fluorophenyl)maleimide 98.0+%, TCI America™

Catalog No. F1054200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1054200MG 200 mg
F10541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F1054200MG Supplier TCI America Supplier No. F1054200MG

Chemical Identifiers

CAS 6633-22-3
Molecular Formula C10H6FNO2
Molecular Weight (g/mol) 191.16
MDL Number MFCD00033653
InChI Key SBKKXWSZVVDOLR-UHFFFAOYSA-N
PubChem CID 96153
IUPAC Name 1-(4-fluorophenyl)-2,5-dihydro-1H-pyrrole-2,5-dione
SMILES FC1=CC=C(C=C1)N1C(=O)C=CC1=O

Specifications

Melting Point 156°C
Color Yellow
Quantity 200 mg
Formula Weight 191.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Fluorophenyl)maleimide