accessibility menu, dialog, popup

UserName

CAS RN 6633-22-3

FNOO

CAS RN 6633-22-3

IUPAC Name: 1-(4-fluorophenyl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-(4-fluorophenyl)pyrrole-2,5-dione
Molecular Weight (g/mol): 191.16
Molecular Formula: C10H6FNO2
InChi Key: SBKKXWSZVVDOLR-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)N1C(=O)C=CC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results