missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Fluorophenyl)formamide 98.0+%, TCI America™

Catalog No. F05465G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F05465G Supplier TCI America Supplier No. F05465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 459-25-6
Molecular Formula C7H6FNO
Molecular Weight (g/mol) 139.129
InChI Key BUPDLPLGFRDHSJ-UHFFFAOYSA-N
Synonym 1-Fluoro-4-formamidobenzene, 4′C-Fluoroformanilide
PubChem CID 237239
IUPAC Name N-(4-fluorophenyl)formamide
SMILES C1=CC(=CC=C1NC=O)F

Specifications

Melting Point 67°C
Quantity 5 g
Formula Weight 139.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Fluorophenyl)formamide