accessibility menu, dialog, popup

UserName

FNHO

CAS RN 459-25-6

IUPAC Name: N-(4-fluorophenyl)formamide
Synonyms: P-fluoroformanilide
Molecular Weight (g/mol): 139.13
Molecular Formula: C7H6FNO
InChi Key: BUPDLPLGFRDHSJ-UHFFFAOYSA-N
SMILES: FC1=CC=C(NC=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results