missing translation for 'onlineSavingsMsg'
Learn More

N,4-Dimethoxy-N-methylbenzamide 95.0+%, TCI America™

Catalog No. D54751G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D54751G 1 g
D54755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D54751G Supplier TCI America Supplier No. D54751G
Only null left

Chemical Identifiers

CAS 52898-49-4
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.218
MDL Number MFCD02684314
InChI Key FIRHLPVYIMMZPV-UHFFFAOYSA-N
PubChem CID 15105748
IUPAC Name N,4-dimethoxy-N-methylbenzamide
SMILES CN(C(=O)C1=CC=C(C=C1)OC)OC

Specifications

Color Yellow
Boiling Point 105°C
Quantity 1 g
Formula Weight 195.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material N,4-Dimethoxy-N-methylbenzamide