missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Cyanophenyl)glycine 98.0+%, TCI America™

Catalog No. c277125g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C277125G 25 g
C27715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C277125G Supplier TCI America Supplier No. C277125G
Only null left

Chemical Identifiers

CAS 42288-26-6
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
MDL Number MFCD01464022
InChI Key KJRQMXRCZULRHF-UHFFFAOYSA-N
PubChem CID 817965
IUPAC Name 2-(4-cyanoanilino)acetic acid
SMILES C1=CC(=CC=C1C#N)NCC(=O)O

Specifications

Melting Point 237°C
Color White-Yellow
Quantity 25 g
Formula Weight 176.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Cyanophenyl)glycine