accessibility menu, dialog, popup

UserName

NHNOHO

CAS RN 42288-26-6

IUPAC Name: 2-[(4-cyanophenyl)amino]acetic acid
Synonyms: [(4-cyanophenyl)amino]acetic acid
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: KJRQMXRCZULRHF-UHFFFAOYSA-N
SMILES: OC(=O)CNC1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results