missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Cyanophenyl)glycine 98.0+%, TCI America™

Catalog No. C27715G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27715G 5 g
C277125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27715G Supplier TCI America Supplier No. C27715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 42288-26-6
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
MDL Number MFCD01464022
InChI Key KJRQMXRCZULRHF-UHFFFAOYSA-N
PubChem CID 817965
IUPAC Name 2-(4-cyanoanilino)acetic acid
SMILES C1=CC(=CC=C1C#N)NCC(=O)O

Specifications

Melting Point 237°C
Color White-Yellow
Quantity 5 g
Formula Weight 176.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Cyanophenyl)glycine