missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[4-Cyano-3-(trifluoromethyl)phenyl]methacrylamide 98.0+%, TCI America™
$139.81 - $419.41
Chemical Identifiers
| CAS | 90357-53-2 |
|---|---|
| Molecular Formula | C12H9F3N2O |
| Molecular Weight (g/mol) | 254.212 |
| MDL Number | MFCD03411609 |
| InChI Key | HHWDZLSGDDXUSM-UHFFFAOYSA-N |
| PubChem CID | 11149469 |
| IUPAC Name | N-[4-cyano-3-(trifluoromethyl)phenyl]-2-methylprop-2-enamide |
| SMILES | CC(=C)C(=O)NC1=CC(=C(C=C1)C#N)C(F)(F)F |
Chemical Identifiers
| 90357-53-2 | |
| 254.212 | |
| HHWDZLSGDDXUSM-UHFFFAOYSA-N | |
| N-[4-cyano-3-(trifluoromethyl)phenyl]-2-methylprop-2-enamide |
| C12H9F3N2O | |
| MFCD03411609 | |
| 11149469 | |
| CC(=C)C(=O)NC1=CC(=C(C=C1)C#N)C(F)(F)F |
Specifications
| 90357-53-2 | |
| White | |
| MFCD03411609 | |
| 3077 | |
| CC(=C)C(=O)NC1=CC(=C(C=C1)C#N)C(F)(F)F | |
| 254.212 | |
| 254.21 | |
| Crystalline Powder |
| 143°C | |
| C12H9F3N2O | |
| 1 g | |
| HHWDZLSGDDXUSM-UHFFFAOYSA-N | |
| N-[4-cyano-3-(trifluoromethyl)phenyl]-2-methylprop-2-enamide | |
| 11149469 | |
| ≥98.0% (GC) | |
| N-[4-Cyano-3-(trifluoromethyl)phenyl]methacrylamide |
Safety and Handling
TSCA : No