missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromophenyl)-N-phenyl-1-naphthylamine 98.0+%, TCI America™

Catalog No. B4299200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4299200MG 200 mg
B42991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4299200MG Supplier TCI America Supplier No. B4299200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 138310-84-6
Molecular Formula C22H16BrN
Molecular Weight (g/mol) 374.281
MDL Number MFCD11046471
InChI Key ABMCIJZTMPDEGW-UHFFFAOYSA-N
Synonym 1-[N-(4-Bromophenyl)anilino]naphthalene
PubChem CID 18398525
IUPAC Name N-(4-bromophenyl)-N-phenylnaphthalen-1-amine
SMILES C1=CC=C(C=C1)N(C2=CC=C(C=C2)Br)C3=CC=CC4=CC=CC=C43

Specifications

Melting Point 93°C
Color White
Quantity 200 mg
Formula Weight 374.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Bromophenyl)-N-phenyl-1-naphthylamine