accessibility menu, dialog, popup

UserName

CAS RN 138310-84-6

IUPAC Name: N-(4-bromophenyl)-N-phenylnaphthalen-1-amine
Synonyms: N-(4-bromophenyl)-N-phenylnaphthalen-1-amine
Molecular Weight (g/mol): 374.28
Molecular Formula: C22H16BrN
InChi Key: ABMCIJZTMPDEGW-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results