missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromobutyl)phthalimide, 98%

Catalog No. p-3735705
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC146290100 10 g Glass bottle
AC146290500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC146290100 Supplier Thermo Scientific Chemicals Supplier No. 146290100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5394-18-3 | C12H12BrNO2 | 282.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5394-18-3
Molecular Formula C12H12BrNO2
Molecular Weight (g/mol) 282.14
MDL Number MFCD00005905
InChI Key UXFWTIGUWHJKDD-UHFFFAOYSA-N
Synonym n-4-bromobutyl phthalimide, 2-4-bromobutyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-4-bromobutyl, 2-4-bromobutyl-1h-isoindole-1,3 2h-dione, n-4-bromobutyl-phthalimide, 2-4-bromobutyl isoindole-1,3-dione, 2-4-bromobutyl-2,3-dihydro-1h-isoindole-1,3-dione, n-4-bromobutyl, n-bromobutylphthalimide
PubChem CID 93575
IUPAC Name 2-(4-bromobutyl)isoindole-1,3-dione
SMILES BrCCCCN1C(=O)C2=CC=CC=C2C1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 76.0°C to 80.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 21, 463
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 282.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-(4-Bromobutyl)phthalimide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 226-401-2

RUO – Research Use Only