accessibility menu, dialog, popup

UserName

CAS RN 5394-18-3

IUPAC Name: 2-(4-bromobutyl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: N-(4-bromobutyl)phthalimide
Molecular Weight (g/mol): 282.14
Molecular Formula: C12H12BrNO2
InChi Key: UXFWTIGUWHJKDD-UHFFFAOYSA-N
SMILES: BrCCCCN1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)